Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Homer2
Entrez Gene ID
26557
Gene Name
homer homolog 2 (Drosophila)
Gene Aliases
9330120H11Rik, AW539445, CPD, Vesl-2
Chromosome Location
Chr.7: 81600481 - 81706925 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164086.1 NP_001157558.1 9 - 9 108
NM_001164087.1 NP_001157559.1 9 - 9 108
NM_011983.2 NP_036113.1 9 - 9 108
GenBank mRNA AA790874.1 - - 108
AK033939.1 - - 108
AK085081.1 - - 108
BG076134.1 - - 640 108

View Full Product Details