Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hba-a1
Entrez Gene ID
15122
Gene Name
hemoglobin alpha, adult chain 1
Gene Aliases
Hba, Hba1, Hbat1
Chromosome Location
Chr.11: 32283672 - 32284486 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008218.2 NP_032244.2 1 - 2 131 94
GenBank mRNA AK003077.1 - 1 - 2 129 94
AK010923.1 - 1 - 2 129 94
AK010927.1 - 1 - 2 130 94
AK010933.1 - 1 - 2 129 94
AK010937.1 - 1 - 2 132 94
AK010987.1 - 1 - 2 129 94
AK011018.1 - 1 - 2 131 94
AK011023.1 - 1 - 2 128 94
AK011025.1 - 1 - 2 131 94
AK011028.1 - 1 - 2 128 94
AK011055.1 - 1 - 2 132 94
AK011063.1 - 1 - 2 136 94
AK011076.1 - 1 - 2 129 94
AK011116.1 - - 642 94
AK012329.1 - 1 - 2 129 94
AK012755.1 - 1 - 2 129 94
AK171893.1 - 1 - 2 129 94
BC043020.1 - 1 - 2 111 94

Gene Symbol
Hba-a2
Entrez Gene ID
110257
Gene Name
hemoglobin alpha, adult chain 2
Gene Aliases
-
Chromosome Location
Chr.11: 32296492 - 32297310 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083955.1 NP_001077424.1 - 128 94
GenBank mRNA AK005434.1 - - 129 94
AK010422.1 - - 129 94
AK011030.1 - - 129 94
AK011073.1 - - 129 94
AK011121.1 - - 129 94
AK012348.1 - - 129 94
AK019138.1 - - 130 94
AK028173.1 - - 129 94
AK076068.1 - - 129 94
CF740193.1 - - 115 94
M26895.1 - - 102 94

View Full Product Details