Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ube3a
Entrez Gene ID
22215
Gene Name
ubiquitin protein ligase E3A
Gene Aliases
4732496B02, 5830462N02Rik, A130086L21Rik, Hpve6a, mKIAA4216
Chromosome Location
Chr.7: 59227814 - 59311536 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033962.1 NP_001029134.1 9 - 10 2847 101
NM_011668.2 NP_035798.2 11 - 12 3034 101
GenBank mRNA AK006957.1 - 2 - 3 228 101
AK192185.1 - 1 - 2 76 101
AK220549.1 - 9 - 10 2747 101
U82122.1 - 7 - 8 2284 101

View Full Product Details