Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Akr1b3
- Entrez Gene ID
-
11677
- Gene Name
- aldo-keto reductase family 1, member B3 (aldose reductase)
- Gene Aliases
-
ALR2, AR, Ahr-1, Ahr1, Akr1b1, Aldor1, Aldr1
- Chromosome Location
-
Chr.6: 34303930 - 34317489 on Build GRCm38
- Gene Symbol
- Gm6644
- Entrez Gene ID
-
626009
- Gene Name
- Akr1b3 pseudogene
- Gene Aliases
-
Akr1b3p, EG626009
- Chromosome Location
-
Chr.1: 55443695 - 55445063 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037965.1 |
|
1 - 1 |
861 |
77 |
View Full Product Details