Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Peg13
Entrez Gene ID
353342
Gene Name
paternally expressed 13
Gene Aliases
-
Chromosome Location
Chr.15: 72805604 - 72810324 on Build GRCm38
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location Amplicon Length
RefSeq NR_002864.1
1 - 1 157 62
GenBank mRNA AY151252.1 - LIT1829
Peg13
1 - 1 157 62


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY248760.1 - 1 139 Chr. 15 - 72809851 - 72810306 [-] on Build GRCm38 62
GenBankmRNA BY230204.1 - 1 148 Chr. 15 - 72809951 - 72810314 [-] on Build GRCm38 62
GenBankmRNA BY234523.1 - 1 148 Chr. 15 - 72809965 - 72810314 [-] on Build GRCm38 62


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream potassium channel, subfamily K, member 9 Kcnk9 Task3 -257.7kb Sense Chr. - 72502927 - 72547853 on Build GRCm38
Downstream chromatin accessibility complex 1 Chrac1 2410152E03Rik, 2810406L04Rik, YCL1 280.1kb Anti-sense Chr. - 73090412 - 73094075 on Build GRCm38

View Full Product Details