The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128093.1 | NP_001121565.1 | 1 - 2 | 205 | 82 |
GenBank mRNA | BC147762.1 | - | 1 - 2 | 205 | 82 |
BC147764.1 | - | 1 - 2 | 205 | 82 |