Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- C1galt1
- Entrez Gene ID
-
94192
- Gene Name
- core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1
- Gene Aliases
-
2210410E06Rik, AV284120, T-synthase
- Chromosome Location
-
Chr.6: 7845224 - 7872042 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK033924.1 |
- |
- |
1451 |
101 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CF173442.1 |
- |
1 |
630 |
|
Chr. 6 - 7872137 - 7872815 [+] on Build GRCm38 |
101 |
GenBankmRNA |
CD806735.1 |
- |
1 |
287 |
|
Chr. 6 - 7872480 - 7873111 [+] on Build GRCm38 |
101 |
GenBankmRNA |
CF583949.1 |
- |
|
565 |
|
Chr. - - [-] on Build |
101 |
GenBankmRNA |
AI785082.1 |
- |
1 |
92 |
|
Chr. 6 - 7872675 - 7873000 [+] on Build GRCm38 |
101 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Downstream |
collagen, type XXVIII, alpha 1 |
Col28a1 |
Col28, Gm466 |
124.0kb
|
Anti-sense |
Chr. - 7997100 - 8194223 on Build GRCm38 |
View Full Product Details