The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027054.1 | NP_081330.1 | 2 - 3 | 341 | 99 |
GenBank mRNA | AF440737.2 | - | 2 - 3 | 323 | 99 |
AK005670.1 | - | 2 - 3 | 332 | 99 | |
BC119432.2 | - | 2 - 3 | 341 | 99 |