The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010916.2 | NP_035046.1 | 1 - 2 | 308 | 66 |
GenBank mRNA | AK149315.1 | - | 1 - 2 | 309 | 66 |
BC051018.1 | - | 1 - 2 | 283 | 66 | |
BC054760.1 | - | 1 - 2 | 284 | 66 |