Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nudt16l1
Entrez Gene ID
66911
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 16-like 1
Gene Aliases
1110001K21Rik, 5330437I08Rik, Sdos
Chromosome Location
Chr.16: 4939111 - 4941021 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025839.3 NP_080115.1 1 - 2 183 88
GenBank mRNA AF435792.1 - 1 - 2 195 88
AK002378.1 - 1 - 2 176 88
AK003241.1 - 1 - 2 184 88
AK007897.1 - 1 - 2 179 88
AK019936.1 - 1 - 2 186 88
AK159841.1 - 1 - 2 176 88
BC025569.1 - 1 - 2 182 88
BC028494.1 - 1 - 2 176 88

View Full Product Details