The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053079.2 | NP_444309.2 | 22 - 23 | 1971 | 98 |
GenBank mRNA | AF205540.1 | - | 22 - 23 | 1970 | 98 |
AK136428.1 | - | 22 - 23 | 1972 | 98 | |
BC116248.1 | - | 22 - 23 | 1961 | 98 | |
BC116249.1 | - | 22 - 23 | 1961 | 98 |