The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177718.3 | NP_808386.2 | 1 - 2 | 245 | 110 |
GenBank mRNA | BC068245.1 | - | 1 - 2 | 233 | 110 |
BC085092.1 | - | 1 - 2 | 171 | 110 | |
BE983254.1 | - | 1 - 2 | 245 | 110 |