Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Usp17ld
- Entrez Gene ID
-
384701
- Gene Name
- ubiquitin specific peptidase 17-like D
- Gene Aliases
-
Dub2a, Dub4
- Chromosome Location
-
Chr.7: 103250086 - 103252505 on Build GRCm38
- Gene Symbol
- Usp17lc
- Entrez Gene ID
-
13532
- Gene Name
- ubiquitin specific peptidase 17-like C
- Gene Aliases
-
Dub-2, Dub2, Dub2b, Usp17l5
- Chromosome Location
-
Chr.7: 103416696 - 103419174 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_010089.3 |
NP_034219.3 |
- |
48 |
58 |
View Full Product Details