Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Speer8-ps1
- Entrez Gene ID
-
74062
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 8, pseudogene 1
- Gene Aliases
-
4933405L09Rik, SPEER-8, Speer8-ps
- Chromosome Location
-
Chr.5: 14945294 - 14978541 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_001584.3 |
|
1 - 2 |
49 |
80 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
spermatogenesis associated glutamate (E)-rich protein 4e |
Speer4e |
|
-6.8kb
|
Anti-sense |
Chr. - 14933631 - 14938475 on Build GRCm38 |
Downstream |
predicted gene 10354 |
Gm10354 |
ENSMUSG00000072188 |
13.9kb
|
Anti-sense |
Chr. - 14974144 - 14979490 on Build GRCm38 |
View Full Product Details