The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244763.1 | NP_001231692.1 | 2 - 3 | 198 | 161 |
GenBank mRNA | AB306997.1 | - | 2 - 3 | 198 | 161 |