Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SOSTDC1
Entrez Gene ID
101104056
Gene Name
sclerostin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.4: 28431747 - 28436037 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007767.4 XP_004007816.1 1 - 2 344 65

View Full Product Details