Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDK2AP1
Entrez Gene ID
101122554
Gene Name
cyclin dependent kinase 2 associated protein 1
Gene Aliases
-
Chromosome Location
Chr.17: 61295202 - 61305636 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027956427.1 XP_027812228.1 3 - 4 548 63

View Full Product Details