Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MANEAL
Entrez Gene ID
101115836
Gene Name
mannosidase endo-alpha like
Gene Aliases
-
Chromosome Location
Chr.1: 13051403 - 13059618 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968103.1 XP_027823904.1 2 - 3 656 70

View Full Product Details