Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DHRS9
Entrez Gene ID
101120807
Gene Name
dehydrogenase/reductase 9
Gene Aliases
-
Chromosome Location
Chr.2: 151390771 - 151410845 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004004622.4 XP_004004671.1 4 - 5 930 94

View Full Product Details