Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R11
Entrez Gene ID
101112519
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 11
Gene Aliases
-
Chromosome Location
Chr.20: 30856282 - 30860122 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019006.4 XP_004019055.1 1 - 2 283 72

View Full Product Details