Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2H2orf72
Entrez Gene ID
101121755
Gene Name
chromosome 2 C2orf72 homolog
Gene Aliases
-
Chromosome Location
Chr.2: 248468577 - 248478095 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015093595.2 XP_014949081.2 2 - 3 715 111

View Full Product Details