Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INHBE
Entrez Gene ID
101121588
Gene Name
inhibin subunit beta E
Gene Aliases
-
Chromosome Location
Chr.3: 173812834 - 173815299 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006544.4 XP_004006593.1 1 - 2 711 81

View Full Product Details