Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
WDR86
Entrez Gene ID
101115118
Gene Name
WD repeat domain 86
Gene Aliases
-
Chromosome Location
Chr.4: 124000139 - 124025453 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969028.1 XP_027824829.1 5 - 6 1225 80
XM_027969029.1 XP_027824830.1 5 - 6 814 80

View Full Product Details