Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A9
Entrez Gene ID
101120546
Gene Name
solute carrier family 16 member 9
Gene Aliases
-
Chromosome Location
Chr.25: 15087269 - 15134698 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962235.1 XP_027818036.1 2 - 3 484 107

View Full Product Details