Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC101119638
Entrez Gene ID
101119638
Gene Name
podocan-like protein 1
Gene Aliases
PODNL1
Chromosome Location
Chr.5: 10097792 - 10102579 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969510.1 XP_027825311.1 3 - 4 509 92

View Full Product Details