Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRIT3
Entrez Gene ID
101118529
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 3
Gene Aliases
-
Chromosome Location
Chr.6: 18669626 - 18685325 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015096376.2 XP_014951862.2 3 - 4 900 86

View Full Product Details