Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIPPLY1
Entrez Gene ID
101116129
Gene Name
ripply transcriptional repressor 1
Gene Aliases
-
Chromosome Location
Chr.X: 136869445 - 136872784 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963056.1 XP_027818857.1 3 - 4 472 69
XM_027963057.1 XP_027818858.1 3 - 4 451 69

View Full Product Details