Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EGLN2
Entrez Gene ID
101120517
Gene Name
egl-9 family hypoxia inducible factor 2
Gene Aliases
-
Chromosome Location
Chr.14: 52645170 - 52654348 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978291.1 XP_027834092.1 5 - 6 1476 67

View Full Product Details