Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FSCN1
Entrez Gene ID
101108186
Gene Name
fascin actin-bundling protein 1
Gene Aliases
-
Chromosome Location
Chr.24: 39569759 - 39579242 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027961638.1 XP_027817439.1 1 - 2 956 68

View Full Product Details