Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MGP
Entrez Gene ID
101106925
Gene Name
matrix Gla protein
Gene Aliases
-
Chromosome Location
Chr.3: 215145542 - 215149123 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006833.3 XP_004006882.1 3 - 4 247 84

View Full Product Details