Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLX4
Entrez Gene ID
101123086
Gene Name
SLX4 structure-specific endonuclease subunit
Gene Aliases
-
Chromosome Location
Chr.24: 2999764 - 3018895 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027961296.1 XP_027817097.1 7 - 8 1847 148

View Full Product Details