Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CENPV
Entrez Gene ID
105611619
Gene Name
centromere protein V
Gene Aliases
-
Chromosome Location
Chr.11: 30013852 - 30021902 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027974842.1 XP_027830643.1 4 - 5 635 98

View Full Product Details