Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR6
Entrez Gene ID
101108523
Gene Name
WD repeat domain 6
Gene Aliases
-
Chromosome Location
Chr.19: 52954421 - 52962948 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012100081.3 XP_011955471.2 5 - 6 3036 98
XM_012100082.3 XP_011955472.2 5 - 6 3036 98

View Full Product Details