Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRIF1
Entrez Gene ID
101105972
Gene Name
ligand dependent nuclear receptor interacting factor 1
Gene Aliases
-
Chromosome Location
Chr.1: 92896810 - 92917485 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012178334.3 XP_012033724.1 2 - 3 1949 118

View Full Product Details