Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NREP
Entrez Gene ID
101105161
Gene Name
neuronal regeneration related protein
Gene Aliases
-
Chromosome Location
Chr.7: 2189181 - 2221588 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010176.4 XP_004010225.1 3 - 4 255 92

View Full Product Details