Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL18
Entrez Gene ID
101111291
Gene Name
mitochondrial ribosomal protein L18
Gene Aliases
-
Chromosome Location
Chr.8: 90783120 - 90788514 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011437.4 XP_004011486.2 2 - 3 355 105

View Full Product Details