Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AXDND1
Entrez Gene ID
101118888
Gene Name
axonemal dynein light chain domain containing 1
Gene Aliases
-
Chromosome Location
Chr.12: 65003699 - 65079785 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004014094.4 XP_004014143.1 13 - 14 1483 112

View Full Product Details