Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPATA12
Entrez Gene ID
105603624
Gene Name
spermatogenesis associated 12
Gene Aliases
-
Chromosome Location
Chr.19: 45995333 - 45997059 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957893.1 XP_027813694.1 2 - 2 418 91

View Full Product Details