Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CA12
Entrez Gene ID
100008777
Gene Name
carbonic anhydrase 12
Gene Aliases
CAR12
Chromosome Location
Chr.17: 8882935 - 8930581 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082086.1 NP_001075555.1 3 - 4 286 75
XM_017347899.1 XP_017203388.1 3 - 4 422 75
GenBank mRNA AF263367.2 - 3 - 4 286 75

View Full Product Details