Gene Transcripts

Gene Symbol
Cyp4f4
Entrez Gene ID
286904
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 4
Gene Aliases
Chromosome Location
Chr. 7: 14891062 - 14907250 on Build Rnor_6.0
UniGene ID
Rn.10170
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173123.1 NP_775146.1 10, 11 1399
GenBank mRNA U39206.1 - 10, 11 1399

View Full Product Details