Gene Transcripts

Gene Symbol
Zbtb11
Entrez Gene ID
304010
Gene Name
zinc finger and BTB domain containing 11
Gene Aliases
Chromosome Location
Chr. 11: 47079201 - 47113993 on Build Rnor_6.0
UniGene ID
Rn.162011
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107097.1 NP_001100567.1 6 1982
XM_006248251.2 XP_006248313.1 6 1658

View Full Product Details