Gene Transcripts

Gene Symbol
Eml4
Entrez Gene ID
313861
Gene Name
echinoderm microtubule associated protein like 4
Gene Aliases
Chromosome Location
Chr. 6: 6618369 - 6674550 on Build Rnor_6.0
UniGene ID
Rn.22926
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108008.1 NP_001101478.1 19, 20 2071
XM_006239681.2 XP_006239743.1 20, 21 2958
XM_006239682.2 XP_006239744.1 19, 20 2925

View Full Product Details