Gene Transcripts

Gene Symbol
Sftpd
Entrez Gene ID
25350
Gene Name
surfactant protein D
Gene Aliases
SP-D, SPD
Chromosome Location
Chr. 16: 18753456 - 18766100 on Build Rnor_6.0
UniGene ID
Rn.11348
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012878.2 NP_037010.1 3 357
XM_008771065.1 XP_008769287.1 3 360
GenBank mRNA BC070507.1 - 4 366
M81231.1 - 4 364

View Full Product Details