Gene Transcripts

Gene Symbol
Ngdn
Entrez Gene ID
305887
Gene Name
neuroguidin, EIF4E binding protein
Gene Aliases
RGD1310211
Chromosome Location
Chr. 15: 33676612 - 33684840 on Build Rnor_6.0
UniGene ID
Rn.228994
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107263.1 NP_001100733.1 6, 7 461
XM_006251966.2 XP_006252028.1 6, 7 654
XM_008770676.1 XP_008768898.1 6, 7 1446

View Full Product Details