Gene Transcripts

Gene Symbol
Cts8l1
Entrez Gene ID
290972
Gene Name
cathepsin 8-like 1
Gene Aliases
Cts8
Chromosome Location
Chr. 17: 3563190 - 3570829 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_225128.8 XP_225128.7 3 333

View Full Product Details