Gene Transcripts

Gene Symbol
Ctxn1
Entrez Gene ID
29145
Gene Name
cortexin 1
Gene Aliases
Ctxn
Chromosome Location
Chr. 12: 2534212 - 2535823 on Build Rnor_6.0
UniGene ID
Rn.9131
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109935.1 NP_001103405.1 2 437
GenBank mRNA DY309362.1 - 2 383
L15011.1 - 2 436

View Full Product Details