Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014061.1 | NP_001014083.1 | 808 | ||
GenBank mRNA | BC079135.1 | - | 808 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_006237686.2 | XP_006237748.1 | 6 | 1213 | |
XM_008763455.1 | XP_008761677.1 | 7 | 2221 | ||
XM_008763456.1 | XP_008761678.1 | 7 | 3595 |