Gene Transcripts

Gene Symbol
Rnf113a1
Entrez Gene ID
313450
Gene Name
ring finger protein 113A1
Gene Aliases
Chromosome Location
Chr. X: 124516953 - 124518080 on Build Rnor_6.0
UniGene ID
Rn.34457
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014791.1 NP_001014791.1 436
GenBank mRNA AY363108.1 - 440
BC087595.1 - 436 7310345

Gene Symbol
Rnf113a
Entrez Gene ID
103694508
Gene Name
ring finger protein 113A
Gene Aliases
Chromosome Location
Chr. X: 123806900 - 123808049 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773490.1 XP_008771712.1 1 458

View Full Product Details