Gene Transcripts

Gene Symbol
Syne1
Entrez Gene ID
499010
Gene Name
spectrin repeat containing, nuclear envelope 1
Gene Aliases
CPG2, Myne-1
Chromosome Location
Chr. 1: 41608287 - 41763602 on Build Rnor_6.0
UniGene ID
Rn.226002
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006227851.2 XP_006227913.1 5089
XM_008758713.1 XP_008756935.1 5104
GenBank mRNA AF452647.1 - 4483

View Full Product Details