Gene Transcripts

Gene Symbol
Vom2r25
Entrez Gene ID
502286
Gene Name
vomeronasal 2 receptor, 25
Gene Aliases
RGD1564853
Chromosome Location
Chr. 8: 3839028 - 3882077 on Build Rnor_6.0
UniGene ID
Rn.217948
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099482.1 NP_001092952.1 6 2285

Gene Symbol
LOC103689970
Entrez Gene ID
103689970
Gene Name
vomeronasal type-2 receptor 116-like
Gene Aliases
Chromosome Location
Chr. 8: 4167412 - 4195297 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008765901.1 XP_008764123.1 4 1043

View Full Product Details